The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017922.1 | NP_001017922.1 | 10 - 11 | 955 | 67 |
NM_018538.3 | NP_061008.2 | 9 - 10 | 851 | 67 | |
XM_006710313.3 | XP_006710376.1 | 11 - 12 | 1220 | 67 | |
XM_011540570.2 | XP_011538872.1 | 10 - 11 | 911 | 67 | |
GenBank mRNA | AF311285.1 | - | 9 - 10 | 851 | 67 |
AK056138.1 | - | 10 - 11 | 929 | 67 | |
AK125447.1 | - | 9 - 10 | 699 | 67 | |
AK314436.1 | - | 10 - 11 | 939 | 67 | |
AY049028.1 | - | 9 - 10 | 869 | 67 | |
BC099703.1 | - | 9 - 10 | 784 | 67 | |
BC099707.1 | - | 9 - 10 | 789 | 67 | |
BC099712.1 | - | 9 - 10 | 784 | 67 | |
BC099713.1 | - | 9 - 10 | 784 | 67 | |
BX537371.1 | - | 7 - 8 | 1453 | 67 |