The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286069.1 | NP_001272998.1 | 1 - 2 | 127 | 62 |
NM_001286071.1 | NP_001273000.1 | 1 - 2 | 127 | 62 | |
NM_014039.3 | NP_054758.2 | 1 - 2 | 127 | 62 | |
GenBank mRNA | AK291833.1 | - | 1 - 2 | 33 | 62 |
AL136605.1 | - | 1 - 2 | 53 | 62 | |
AL708049.1 | - | 1 - 2 | 134 | 62 | |
BC012298.1 | - | 1 - 2 | 94 | 62 | |
BM551310.1 | - | 1 - 2 | 79 | 62 |