Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TAF1D
- Entrez Gene ID
-
79101
- Gene Name
- TATA-box binding protein associated factor, RNA polymerase I subunit D
- Gene Aliases
-
JOSD3, RAFI41, TAF(I)41, TAFI41
- Chromosome Location
-
Chr.11: 93735929 - 93741537 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK027016.1 |
- |
7 - 8 |
1101 |
121 |
View Full Product Details