The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000615.6 | NP_000606.3 | 10 - 11 | 1762 | 109 |
NM_001076682.3 | NP_001070150.1 | 10 - 11 | 1762 | 109 | |
NM_001242607.1 | NP_001229536.1 | 12 - 13 | 1870 | 109 | |
NM_001242608.1 | NP_001229537.1 | 10 - 11 | 1762 | 109 | |
NM_181351.4 | NP_851996.2 | 11 - 12 | 1792 | 109 | |
GenBank mRNA | AB209443.1 | - | 10 - 11 | 1641 | 109 |
AK057509.1 | - | 10 - 11 | 1865 | 109 | |
AK292453.1 | - | 11 - 12 | 1427 | 109 | |
AK314589.1 | - | 12 - 13 | 1842 | 109 | |
BC014205.2 | - | 5 - 6 | 656 | 109 | |
BC047244.1 | - | 11 - 12 | 1604 | 109 | |
S71824.1 | - | 10 - 11 | 1597 | 109 | |
U63041.1 | - | 10 - 11 | 1433 | 109 | |
X16841.1 | - | 10 - 11 | 1541 | 109 |