The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000615.6 | NP_000606.3 | 1 - 2 | 413 | 84 |
NM_001076682.3 | NP_001070150.1 | 1 - 2 | 413 | 84 | |
NM_001242607.1 | NP_001229536.1 | 1 - 2 | 413 | 84 | |
NM_001242608.1 | NP_001229537.1 | 1 - 2 | 413 | 84 | |
NM_181351.4 | NP_851996.2 | 1 - 2 | 413 | 84 | |
GenBank mRNA | AB209443.1 | - | 1 - 2 | 292 | 84 |
AF005070.1 | - | 1 - 2 | 259 | 84 | |
AK292453.1 | - | 1 - 2 | 48 | 84 | |
AK314589.1 | - | 1 - 2 | 385 | 84 | |
BC029119.1 | - | 1 - 2 | 224 | 84 | |
BC047244.1 | - | 1 - 2 | 225 | 84 | |
DA337649.1 | - | 1 - 2 | 387 | 84 | |
S71824.1 | - | 1 - 2 | 248 | 84 | |
U63041.1 | - | 1 - 2 | 84 | 84 | |
X16841.1 | - | 1 - 2 | 192 | 84 |