Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VWA5A
Entrez Gene ID
4013
Gene Name
von Willebrand factor A domain containing 5A
Gene Aliases
BCSC-1, BCSC1, LOH11CR2A
Chromosome Location
Chr.11: 124115404 - 124146912 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130142.1 NP_001123614.1 13 - 14 1733 118
NM_014622.4 NP_055437.2 12 - 13 1618 118
XM_011542828.2 XP_011541130.1 13 - 14 1844 118
GenBank mRNA AF002672.1 - 11 - 12 1530 118
AK291698.1 - 12 - 13 1572 118
AK314934.1 - 5 - 6 583 118
AY366505.1 - 12 - 13 1618 118
AY366508.1 - 13 - 14 1733 118

View Full Product Details