Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VWA5A
Entrez Gene ID
4013
Gene Name
von Willebrand factor A domain containing 5A
Gene Aliases
BCSC-1, BCSC1, LOH11CR2A
Chromosome Location
Chr.11: 124115404 - 124146912 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130142.1 NP_001123614.1 16 - 17 2233 89
NM_014622.4 NP_055437.2 15 - 16 2118 89
XM_011542828.2 XP_011541130.1 16 - 17 2344 89
GenBank mRNA AF002672.1 - 14 - 15 2030 89
AK291698.1 - 15 - 16 2072 89
AK314934.1 - 8 - 9 1083 89
AY366505.1 - 15 - 16 2118 89
AY366508.1 - 16 - 17 2233 89

View Full Product Details