The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138294.2 | NP_612151.1 | 1 - 2 | 62 | 144 |
GenBank mRNA | AF462605.1 | - | 1 - 2 | 58 | 144 |
BC039863.1 | - | 1 - 2 | 62 | 144 | |
BC107044.2 | - | 1 - 2 | 58 | 108 |