The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_017002145.1 | XP_016857634.1 | 4 - 5 | 591 | 95 |
XM_017002146.1 | XP_016857635.1 | 4 - 5 | 461 | 95 | |
GenBank mRNA | AK307645.1 | - | 3 - 4 | 403 | 95 |