Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SYNE1
- Entrez Gene ID
-
23345
- Gene Name
- spectrin repeat containing nuclear envelope protein 1
- Gene Aliases
-
8B, ARCA1, C6orf98, CPG2, EDMD4, MYNE1, Nesp1, SCAR8, dJ45H2.2
- Chromosome Location
-
Chr.6: 152121684 - 152637399 on Build GRCh38
View Full Product Details