Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SYNE1
Entrez Gene ID
23345
Gene Name
spectrin repeat containing nuclear envelope protein 1
Gene Aliases
8B, ARCA1, C6orf98, CPG2, EDMD4, MYNE1, Nesp1, SCAR8, dJ45H2.2
Chromosome Location
Chr.6: 152121684 - 152637399 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033071.3 NP_149062.1 26 - 27 3640 69
NM_182961.3 NP_892006.3 26 - 27 3784 69
XM_006715407.1 XP_006715470.1 26 - 27 3682 69
XM_006715408.2 XP_006715471.1 26 - 27 3686 69
XM_006715409.1 XP_006715472.1 25 - 26 3661 69
XM_006715410.2 XP_006715473.1 26 - 27 3686 69
XM_006715411.1 XP_006715474.1 25 - 26 3631 69
XM_006715412.2 XP_006715475.1 26 - 27 3686 69
XM_006715413.2 XP_006715476.1 26 - 27 3686 69
XM_006715414.1 XP_006715477.1 24 - 25 3610 69
XM_006715415.2 XP_006715478.1 26 - 27 3686 69
XM_006715416.2 XP_006715479.1 26 - 27 3686 69
XM_006715417.2 XP_006715480.1 26 - 27 3686 69
XM_006715420.2 XP_006715483.1 26 - 27 3686 69
XM_006715421.2 XP_006715484.1 26 - 27 3686 69
XM_006715422.1 XP_006715485.1 25 - 26 3523 69
XM_006715423.2 XP_006715486.1 26 - 27 3686 69
XM_006715424.2 XP_006715487.1 26 - 27 3686 69
XM_006715425.2 XP_006715488.1 26 - 27 3686 69
XM_011535641.2 XP_011533943.1 26 - 27 3686 69
XM_011535642.2 XP_011533944.1 26 - 27 3686 69
XM_011535643.1 XP_011533945.1 25 - 26 3517 69
XM_011535644.1 XP_011533946.1 12 - 13 1733 69
XM_011535645.2 XP_011533947.1 8 - 9 1126 69
XM_017010608.1 XP_016866097.1 27 - 28 3801 69
XM_017010609.1 XP_016866098.1 26 - 27 3608 69
XM_017010610.1 XP_016866099.1 25 - 26 3587 69
XM_017010611.1 XP_016866100.1 25 - 26 3390 69
XM_017010612.1 XP_016866101.1 24 - 25 3314 69
XM_017010613.1 XP_016866102.1 26 - 27 3686 69
XM_017010614.1 XP_016866103.1 26 - 27 3686 69
XM_017010615.1 XP_016866104.1 26 - 27 3686 69
XM_017010616.1 XP_016866105.1 26 - 27 3686 69
XM_017010617.1 XP_016866106.1 26 - 27 3686 69
XM_017010618.1 XP_016866107.1 26 - 27 3686 69
XM_017010619.1 XP_016866108.1 12 - 13 1765 69
GenBank mRNA AB051543.1 - 13 - 14 1857 69
AF495910.1 - 26 - 27 3784 69
AF535142.1 - 26 - 27 3640 69
AK091651.1 - 2 - 3 270 69
AL713682.1 - 7 - 8 956 69
FM162565.1 - 24 - 25 3400 69
KU310547.1 - 13 - 14 1888 69

View Full Product Details