The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004466.5 | NP_004457.1 | 5 - 6 | 1713 | 70 |
XM_011521054.2 | XP_011519356.1 | 5 - 6 | 1645 | 70 | |
XM_017020435.1 | XP_016875924.1 | 5 - 6 | 1645 | 70 | |
GenBank mRNA | AF001462.1 | - | 5 - 6 | 1366 | 70 |
AK312815.1 | - | 6 - 7 | 1673 | 70 | |
BC039730.1 | - | 6 - 7 | 1675 | 70 | |
U66033.1 | - | 5 - 6 | 1299 | 70 |