The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173560.3 | NP_775831.2 | 10 - 11 | 1078 | 106 |
XM_011535589.1 | XP_011533891.1 | 9 - 10 | 965 | 106 | |
XM_017010477.1 | XP_016865966.1 | 9 - 10 | 885 | 106 | |
GenBank mRNA | BC039248.1 | - | 10 - 11 | 1078 | 106 |