The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173560.3 | NP_775831.2 | 12 - 13 | 1387 | 92 |
XM_011535589.1 | XP_011533891.1 | 11 - 12 | 1274 | 92 | |
XM_017010477.1 | XP_016865966.1 | 11 - 12 | 1194 | 92 | |
GenBank mRNA | BC039248.1 | - | 12 - 13 | 1387 | 92 |