Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CAPN13
Entrez Gene ID
92291
Gene Name
calpain 13
Gene Aliases
-
Chromosome Location
Chr.2: 30610382 - 30809527 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144575.2 NP_653176.2 2 - 3 379 100
XM_011533159.2 XP_011531461.1 2 - 3 376 100
XM_011533160.2 XP_011531462.1 3 - 4 567 100
XM_011533161.2 XP_011531463.1 2 - 3 376 100
XM_011533163.2 XP_011531465.1 2 - 3 376 100
XM_017005264.1 XP_016860753.1 3 - 4 567 100
XM_017005265.1 XP_016860754.1 2 - 3 376 100
XM_017005266.1 XP_016860755.1 2 - 3 376 100
GenBank mRNA AK074418.1 - 3 - 4 446 100
AK290298.1 - 2 - 3 380 100
AK308037.1 - 2 - 3 356 100
BC117345.1 - 2 - 3 340 100
BC117347.1 - 2 - 3 340 100
BX476842.1 - 2 - 3 393 100
BX640793.1 - 2 - 3 379 100

View Full Product Details