Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WFDC10B
Entrez Gene ID
280664
Gene Name
WAP four-disulfide core domain 10B
Gene Aliases
WAP12
Chromosome Location
Chr.20: 45684651 - 45708899 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172131.2 NP_742143.1 2 - 3 163 80
GenBank mRNA AF454507.1 - 2 - 3 163 80

View Full Product Details