The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002535.2 | NP_002526.2 | 4 - 5 | 1000 | 139 |
NM_016817.2 | NP_058197.2 | 4 - 5 | 1000 | 139 | |
GenBank mRNA | AK292796.1 | - | 4 - 5 | 889 | 139 |
AK292906.1 | - | 4 - 5 | 917 | 139 | |
BC049215.1 | - | 4 - 5 | 901 | 139 | |
BQ213828.1 | - | 3 - 4 | 527 | 139 | |
M87284.1 | - | 4 - 5 | 879 | 139 | |
M87434.1 | - | 4 - 5 | 879 | 139 |