Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BUD13
Entrez Gene ID
84811
Gene Name
BUD13 homolog
Gene Aliases
Cwc26, fSAP71
Chromosome Location
Chr.11: 116748170 - 116772998 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032725.3 NP_116114.1 4 - 5 1075 76
XM_011543035.2 XP_011541337.1 4 - 5 1649 76
GenBank mRNA AK289635.1 - 4 - 5 1064 76
AK291111.1 - 4 - 5 1064 76
AK311068.1 - 4 - 5 1064 76
BC006350.2 - 4 - 5 1065 76

View Full Product Details