The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000117.2 | NP_000108.1 | 1 - 2 | 331 | 163 |
GenBank mRNA | BC000738.2 | - | 1 - 2 | 205 | 163 |
BT007401.1 | - | 1 - 2 | 83 | 163 | |
CR536536.1 | - | 1 - 2 | 83 | 163 | |
X82434.1 | - | 1 - 2 | 141 | 163 |