Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SART3
Entrez Gene ID
9733
Gene Name
squamous cell carcinoma antigen recognized by T-cells 3
Gene Aliases
DSAP1, P100, RP11-13G14, TIP110, p110, p110(nrb)
Chromosome Location
Chr.12: 108522214 - 108561402 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014706.3 NP_055521.1 3 - 4 785 105
XM_005269241.4 XP_005269298.1 3 - 4 798 105
XM_011539026.2 XP_011537328.1 2 - 3 238 105
GenBank mRNA AB020880.1 - 3 - 4 569 105
AF387506.1 - 3 - 4 564 105
AK225174.1 - 3 - 4 564 105
AK290209.1 - 3 - 4 577 105
AK299250.1 - 2 - 3 222 105
AK304834.1 - 4 - 5 487 105
BC032601.1 - 3 - 4 568 105
BC093784.1 - 3 - 4 555 105
BC103706.1 - 3 - 4 553 105
BC111983.1 - 3 - 4 555 105
BC143253.1 - 3 - 4 555 105
D63879.1 - 3 - 4 550 105

View Full Product Details