The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005400.2 | NP_005391.1 | 3 - 4 | 767 | 62 |
XM_005264428.1 | XP_005264485.1 | 4 - 5 | 796 | 62 | |
XM_005264431.3 | XP_005264488.1 | 3 - 4 | 1356 | 62 | |
XM_011532971.2 | XP_011531273.1 | 5 - 6 | 1420 | 62 | |
XM_011532975.2 | XP_011531277.1 | 7 - 8 | 908 | 62 | |
XM_011532978.1 | XP_011531280.1 | 4 - 5 | 400 | 62 | |
XM_011532980.2 | XP_011531282.1 | 4 - 5 | 437 | 62 | |
XM_011532981.2 | XP_011531283.1 | 2 - 3 | 1560 | 62 | |
XM_017004486.1 | XP_016859975.1 | 5 - 6 | 1462 | 62 | |
XM_017004487.1 | XP_016859976.1 | 5 - 6 | 509 | 62 | |
XM_017004488.1 | XP_016859977.1 | 4 - 5 | 1244 | 62 | |
XM_017004489.1 | XP_016859978.1 | 8 - 9 | 849 | 62 | |
XM_017004490.1 | XP_016859979.1 | 4 - 5 | 1349 | 62 | |
XM_017004491.1 | XP_016859980.1 | 3 - 4 | 400 | 62 | |
XM_017004492.1 | XP_016859981.1 | 3 - 4 | 1356 | 62 | |
GenBank mRNA | AK313842.1 | - | 3 - 4 | 666 | 62 |
AL832345.1 | - | 3 - 4 | 897 | 62 | |
BC109033.2 | - | 3 - 4 | 705 | 62 | |
BC109034.2 | - | 3 - 4 | 705 | 62 | |
X65293.1 | - | 3 - 4 | 585 | 62 |