The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286574.1 | NP_001273503.1 | 1 - 2 | 245 | 82 |
NM_001286576.1 | NP_001273505.1 | 1 - 2 | 245 | 82 | |
GenBank mRNA | AK058119.1 | - | 1 - 2 | 195 | 82 |
CR735810.1 | - | 1 - 2 | 139 | 82 | |
DB070109.1 | - | 1 - 2 | 245 | 82 |