The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000341.3 | NP_000332.2 | 8 - 9 | 1584 | 66 |
XM_011533047.2 | XP_011531349.1 | 8 - 9 | 1584 | 66 | |
GenBank mRNA | AB033549.1 | - | 8 - 9 | 1565 | 66 |
AK091932.1 | - | 9 - 10 | 1450 | 66 | |
AK092000.1 | - | 5 - 6 | 1690 | 66 | |
AK223146.1 | - | 8 - 9 | 1546 | 66 | |
AK289636.1 | - | 8 - 9 | 1562 | 66 | |
AK309145.1 | - | 8 - 9 | 1578 | 66 | |
AK313157.1 | - | 8 - 9 | 1578 | 66 | |
BC022386.1 | - | 8 - 9 | 1582 | 66 | |
BC093624.1 | - | 8 - 9 | 1543 | 66 | |
BC093626.1 | - | 8 - 9 | 1538 | 66 | |
DQ023512.1 | - | 5 - 6 | 1690 | 66 | |
DQ023514.1 | - | 2 - 3 | 707 | 66 | |
DQ023516.1 | - | 8 - 9 | 1506 | 66 | |
DQ023517.1 | - | 8 - 9 | 1506 | 66 | |
KU178335.1 | - | 8 - 9 | 1506 | 66 | |
L11696.1 | - | 8 - 9 | 1549 | 66 | |
M95548.1 | - | 8 - 9 | 1550 | 66 |