The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_073574.1 | 9 - 10 | 1256 | 113 | |
XM_017025114.1 | XP_016880603.1 | 10 - 11 | 1846 | 113 | |
XM_017025118.1 | XP_016880607.1 | 9 - 10 | 1861 | 113 | |
GenBank mRNA | AB208939.1 | - | 9 - 10 | 1742 | 113 |
AK024672.1 | - | 9 - 10 | 1503 | 113 | |
AK096767.1 | - | 9 - 10 | 1256 | 113 |