The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184797.1 | NP_001171726.1 | 1 - 2 | 166 | 82 |
NM_018196.3 | NP_060666.1 | 1 - 2 | 166 | 82 | |
XM_017029620.1 | XP_016885109.1 | 1 - 2 | 166 | 82 | |
GenBank mRNA | AK001589.1 | - | 1 - 2 | 141 | 82 |
AK291694.1 | - | 1 - 2 | 104 | 82 | |
BC025269.1 | - | 1 - 2 | 82 | 82 | |
DA323339.1 | - | 1 - 2 | 83 | ||
DB174485.1 | - | 1 - 2 | 83 |