Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADAMTS4
Entrez Gene ID
9507
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 4
Gene Aliases
ADAMTS-2, ADAMTS-4, ADMP-1
Chromosome Location
Chr.1: 161189725 - 161199080 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320336.1 NP_001307265.1 2 - 3 1382 64
NM_005099.5 NP_005090.3 2 - 3 1382 64
GenBank mRNA AB014588.1 - 2 - 3 1356 64
AF148213.1 - 2 - 3 1362 64
AK291573.1 - 2 - 3 1381 64
AY358886.1 - 2 - 3 1374 64
BC063293.1 - 2 - 3 1381 64

View Full Product Details