Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
XRN1
Entrez Gene ID
54464
Gene Name
5'-3' exoribonuclease 1
Gene Aliases
SEP1
Chromosome Location
Chr.3: 142306607 - 142448062 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282857.1 NP_001269786.1 7 - 8 907 159
NM_001282859.1 NP_001269788.1 7 - 8 907 159
NM_019001.4 NP_061874.3 7 - 8 907 159
XM_006713673.2 XP_006713736.1 7 - 8 907 159
XM_011512919.2 XP_011511221.1 7 - 8 907 159
XM_011512920.2 XP_011511222.1 7 - 8 907 159
XM_011512922.2 XP_011511224.1 4 - 5 538 159
XM_017006640.1 XP_016862129.1 7 - 8 907 159
XM_017006641.1 XP_016862130.1 7 - 8 907 159
XM_017006642.1 XP_016862131.1 7 - 8 907 159
GenBank mRNA AK096177.1 - 7 - 8 907 159
AK123897.1 - 4 - 5 380 159
AK304312.1 - 4 - 5 538 159
AY137776.1 - 7 - 8 789 159
BC039314.1 - 7 - 8 855 159
BC048104.1 - 7 - 8 814 159
BC078146.1 - 7 - 8 819 159
BX640905.1 - 7 - 8 856 159
CR627396.1 - 7 - 8 859 159
CR749518.1 - 7 - 8 837 159

View Full Product Details