Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR91
Entrez Gene ID
29062
Gene Name
WD repeat domain 91
Gene Aliases
HSPC049, SORF-1, SORF1
Chromosome Location
Chr.7: 135183838 - 135211564 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014149.3 NP_054868.3 1 - 2 184 57
XM_011516083.2 XP_011514385.1 1 - 2 154 57
XM_011516086.2 XP_011514388.1 1 - 2 154 57
XM_017012070.1 XP_016867559.1 1 - 2 154 57
XM_017012072.1 XP_016867561.1 1 - 2 134 57
GenBank mRNA AF161534.1 - 1 - 2 129 57
AK001606.1 - 1 - 2 139 57
AK022935.1 - 1 - 2 128 57
AL136940.1 - 1 - 2 35 57
BC017246.2 - 1 - 2 123 57
BC063394.1 - 1 - 2 120 57
BX640957.1 - 1 - 2 155 57

View Full Product Details