The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006240.2 | NP_006231.2 | 14 - 15 | 1728 | 87 |
NM_152224.1 | NP_689410.1 | 14 - 15 | 1644 | 87 | |
XM_005274553.2 | XP_005274610.1 | 16 - 17 | 2055 | 87 | |
XM_017029612.1 | XP_016885101.1 | 14 - 15 | 1866 | 87 | |
GenBank mRNA | AF023455.1 | - | 11 - 12 | 1329 | 87 |
AF027977.1 | - | 11 - 12 | 1481 | 87 | |
AK290463.1 | - | 11 - 12 | 1383 | 87 | |
AK292235.1 | - | 14 - 15 | 1727 | 87 | |
AK295504.1 | - | 10 - 11 | 1218 | 87 | |
AK300815.1 | - | 10 - 11 | 1246 | 87 | |
AK315809.1 | - | 11 - 12 | 1344 | 87 | |
BC036026.1 | - | 14 - 15 | 1727 | 87 | |
X97867.1 | - | 14 - 15 | 1730 | 87 |