The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198719.1 | NP_001185648.1 | 7 - 8 | 1231 | 144 |
NM_002893.3 | NP_002884.1 | 7 - 8 | 1243 | 144 | |
GenBank mRNA | AK091911.1 | - | 7 - 8 | 1228 | 144 |
AK291567.1 | - | 7 - 8 | 987 | 144 | |
BC114500.1 | - | 7 - 8 | 1141 | 144 | |
BC114501.1 | - | 7 - 8 | 1141 | 144 | |
U35143.1 | - | 7 - 8 | 1146 | 144 | |
X72841.1 | - | 7 - 8 | 1170 | 144 |