The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198719.1 | NP_001185648.1 | 9 - 10 | 1383 | 63 |
NM_002893.3 | NP_002884.1 | 9 - 10 | 1395 | 63 | |
GenBank mRNA | AK091911.1 | - | 9 - 10 | 1380 | 63 |
AK127332.1 | - | 1 - 2 | 104 | 63 | |
AK291567.1 | - | 9 - 10 | 1139 | 63 | |
BC114500.1 | - | 9 - 10 | 1293 | 63 | |
BC114501.1 | - | 9 - 10 | 1293 | 63 | |
CR541701.1 | - | 9 - 10 | 1037 | 63 | |
U35143.1 | - | 9 - 10 | 1298 | 63 | |
X72841.1 | - | 9 - 10 | 1322 | 63 |