The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022144.2 | NP_071427.2 | 3 - 4 | 553 | 146 |
GenBank mRNA | AB055421.1 | - | 3 - 4 | 333 | 146 |
AF191770.1 | - | 3 - 4 | 513 | 146 | |
AF234259.1 | - | 3 - 4 | 394 | 146 | |
AF291656.1 | - | 3 - 4 | 399 | 146 | |
AY358706.1 | - | 3 - 4 | 385 | 146 |