Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LTBP4
Entrez Gene ID
8425
Gene Name
latent transforming growth factor beta binding protein 4
Gene Aliases
ARCL1C, LTBP-4, LTBP4L, LTBP4S
Chromosome Location
Chr.19: 40592888 - 40629820 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042544.1 NP_001036009.1 10 - 11 1356 119
NM_001042545.1 NP_001036010.1 7 - 8 1172 119
NM_003573.2 NP_003564.2 10 - 11 1245 119
XM_011527376.2 XP_011525678.1 10 - 11 1385 119
XM_011527377.2 XP_011525679.1 10 - 11 1404 119
XM_011527378.2 XP_011525680.1 10 - 11 1404 119
XM_011527379.1 XP_011525681.1 7 - 8 1193 119
XM_011527380.2 XP_011525682.1 10 - 11 1404 119
XM_011527381.2 XP_011525683.1 10 - 11 1404 119
XM_011527382.2 XP_011525684.1 10 - 11 1404 119
XM_011527383.2 XP_011525685.1 10 - 11 1404 119
XM_011527384.2 XP_011525686.1 10 - 11 1404 119
XM_011527385.2 XP_011525687.1 10 - 11 1404 119
XM_011527386.2 XP_011525688.1 10 - 11 1404 119
XM_011527387.1 XP_011525689.1 10 - 11 1504 119
XM_017027352.1 XP_016882841.1 10 - 11 1404 119
XM_017027353.1 XP_016882842.1 10 - 11 1404 119
XM_017027354.1 XP_016882843.1 10 - 11 1404 119
GenBank mRNA AF051344.2 - 7 - 8 1172 119
AF051345.2 - 10 - 11 1356 119
Y13622.1 - 9 - 10 1245 119

View Full Product Details