The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042544.1 | NP_001036009.1 | 26 - 27 | 3682 | 104 |
NM_001042545.1 | NP_001036010.1 | 23 - 24 | 3498 | 104 | |
NM_003573.2 | NP_003564.2 | 26 - 27 | 3571 | 104 | |
XM_011527376.2 | XP_011525678.1 | 25 - 26 | 3825 | 104 | |
XM_011527377.2 | XP_011525679.1 | 26 - 27 | 3730 | 104 | |
XM_011527379.1 | XP_011525681.1 | 22 - 23 | 3633 | 104 | |
XM_011527380.2 | XP_011525682.1 | 25 - 26 | 3604 | 104 | |
XM_011527381.2 | XP_011525683.1 | 25 - 26 | 3604 | 104 | |
XM_011527382.2 | XP_011525684.1 | 25 - 26 | 3487 | 104 | |
XM_011527387.1 | XP_011525689.1 | 26 - 27 | 3830 | 104 | |
XM_017027352.1 | XP_016882841.1 | 25 - 26 | 3598 | 104 | |
XM_017027353.1 | XP_016882842.1 | 24 - 25 | 3472 | 104 | |
GenBank mRNA | AF051344.2 | - | 23 - 24 | 3498 | 104 |
AF051345.2 | - | 26 - 27 | 3682 | 104 | |
AF054502.1 | - | 8 - 9 | 1095 | 104 | |
AK074499.1 | - | 4 - 5 | 986 | 104 | |
AK128191.1 | - | 12 - 13 | 1680 | 104 | |
Y13622.1 | - | 25 - 26 | 3571 | 104 |