The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032946.2 | NP_116564.2 | 1 - 2 | 74 | 95 |
NR_028089.1 | 1 - 2 | 74 | 95 | ||
GenBank mRNA | AJ277654.1 | - | 1 - 2 | 74 | 95 |
AJ277655.1 | - | 1 - 2 | 74 | 95 | |
AJ277656.1 | - | 1 - 2 | 74 | 95 | |
AJ277657.1 | - | 1 - 2 | 74 | 95 | |
AJ277658.1 | - | 1 - 2 | 74 | 95 |