The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171184.1 | NP_001164655.1 | 18 - 19 | 2329 | 68 |
NM_001939.2 | NP_001930.2 | 20 - 21 | 2612 | 68 | |
XM_017029333.1 | XP_016884822.1 | 19 - 20 | 2695 | 68 | |
XM_017029334.1 | XP_016884823.1 | 19 - 20 | 2691 | 68 | |
GenBank mRNA | AB209305.1 | - | 11 - 12 | 1209 | 68 |
AK289825.1 | - | 20 - 21 | 2500 | 68 | |
AK295843.1 | - | 18 - 19 | 2198 | 68 | |
BC111695.1 | - | 20 - 21 | 2449 | 68 | |
U43519.1 | - | 20 - 21 | 2561 | 68 |