Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DRP2
Entrez Gene ID
1821
Gene Name
dystrophin related protein 2
Gene Aliases
DRP-2
Chromosome Location
Chr.X: 101219786 - 101264497 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171184.1 NP_001164655.1 6 - 7 1066 67
NM_001939.2 NP_001930.2 8 - 9 1349 67
XM_017029333.1 XP_016884822.1 8 - 9 1507 67
GenBank mRNA AK289825.1 - 8 - 9 1237 67
AK295843.1 - 6 - 7 935 67
BC111695.1 - 8 - 9 1186 67

View Full Product Details