Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DRP2
Entrez Gene ID
1821
Gene Name
dystrophin related protein 2
Gene Aliases
DRP-2
Chromosome Location
Chr.X: 101219786 - 101264497 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171184.1 NP_001164655.1 7 - 8 1143 91
NM_001939.2 NP_001930.2 9 - 10 1426 91
XM_017029333.1 XP_016884822.1 9 - 10 1584 91
GenBank mRNA AK289825.1 - 9 - 10 1314 91
AK295843.1 - 7 - 8 1012 91
BC111695.1 - 9 - 10 1263 91
U43519.1 - 9 - 10 1375 91

View Full Product Details