Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GSTP1
Entrez Gene ID
2950
Gene Name
glutathione S-transferase pi 1
Gene Aliases
DFN7, FAEES3, GST3, GSTP, HEL-S-22, PI
Chromosome Location
Chr.11: 67583595 - 67586653 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000852.3 NP_000843.1 4 - 5 479 75
GenBank mRNA AY579414.1 - 3 - 4 232 75
BC010915.1 - 4 - 5 266 75
BT019949.1 - 3 - 4 230 75
BT019950.1 - 3 - 4 230 75
CA946272.1 - 4 - 5 479 75
CR450361.1 - 3 - 4 230 75
EU794620.1 - 4 - 5 259 75
GQ141553.1 - 1 - 2 62 75
GQ141554.1 - 1 - 2 62 75
U30897.1 - 4 - 5 242 75
U62589.1 - 4 - 5 243 75
X06547.1 - 3 - 4 236 75
X15480.1 - 4 - 5 247 75

View Full Product Details