The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000852.3 | NP_000843.1 | 6 - 7 | 694 | 67 |
GenBank mRNA | BC010915.1 | - | 6 - 7 | 481 | 67 |
BT019949.1 | - | 5 - 6 | 445 | 67 | |
BT019950.1 | - | 5 - 6 | 445 | 67 | |
CR450361.1 | - | 5 - 6 | 445 | 67 | |
EU794620.1 | - | 6 - 7 | 474 | 67 | |
GQ141553.1 | - | 3 - 4 | 277 | 67 | |
GQ141554.1 | - | 3 - 4 | 277 | 67 | |
U30897.1 | - | 6 - 7 | 457 | 67 | |
U62589.1 | - | 6 - 7 | 458 | 67 | |
X06547.1 | - | 5 - 6 | 451 | 67 | |
X15480.1 | - | 6 - 7 | 462 | 67 |