The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017857.3 | NP_060327.3 | 4 - 5 | 642 | 102 |
XM_017017942.1 | XP_016873431.1 | 4 - 5 | 681 | 102 | |
GenBank mRNA | AB072360.3 | - | 4 - 5 | 464 | 102 |
AB099291.1 | - | 4 - 5 | 464 | 102 | |
AK000522.1 | - | 4 - 5 | 610 | 102 | |
AK074432.1 | - | 4 - 5 | 560 | 102 | |
BC007709.1 | - | 4 - 5 | 521 | 102 |