The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271581.1 | NP_001258510.1 | 15 - 16 | 2057 | 151 |
NM_018981.2 | NP_061854.1 | 16 - 17 | 2195 | 151 | |
NR_073365.1 | 16 - 17 | 2949 | 151 | ||
GenBank mRNA | AF038503.1 | - | 16 - 17 | 1906 | 151 |
AF490904.1 | - | 15 - 16 | 1805 | 151 | |
AK027450.1 | - | 14 - 15 | 1536 | 151 | |
AK027647.1 | - | 16 - 17 | 1964 | 151 | |
AK027696.1 | - | 16 - 17 | 1943 | 151 | |
AK027701.2 | - | 13 - 14 | 1317 | 151 | |
AK074905.1 | - | 3 - 4 | 311 | 151 | |
AK298138.1 | - | 11 - 12 | 1042 | 151 | |
AL137648.1 | - | 5 - 6 | 745 | 151 | |
AL832267.1 | - | 9 - 10 | 895 | 151 | |
AL832632.2 | - | 16 - 17 | 2720 | 151 | |
AL832646.3 | - | 14 - 15 | 1546 | 151 | |
AY358577.1 | - | 15 - 16 | 1814 | 151 | |
BC117299.1 | - | 16 - 17 | 1910 | 151 | |
BC126168.1 | - | 16 - 17 | 1910 | 151 |