The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161344.1 | NP_001154816.1 | 2 - 3 | 217 | 77 |
NM_001161345.1 | NP_001154817.1 | 2 - 3 | 217 | 77 | |
NM_001161346.1 | NP_001154818.1 | 2 - 3 | 217 | 77 | |
NM_001161347.1 | NP_001154819.1 | 2 - 3 | 217 | 77 | |
NM_018223.2 | NP_060693.2 | 2 - 3 | 217 | 77 | |
GenBank mRNA | AF170724.1 | - | 2 - 3 | 223 | 77 |
AK001658.1 | - | 2 - 3 | 197 | 77 | |
AK027687.1 | - | 2 - 3 | 211 | 77 | |
AK302785.1 | - | 2 - 3 | 202 | 77 | |
AK304333.1 | - | 2 - 3 | 204 | 77 | |
BC012072.1 | - | 1 - 2 | 141 | 77 | |
BP313700.1 | - | 2 - 3 | 217 | 77 |