The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291494.1 | NP_001278423.1 | 16 - 17 | 2287 | 142 |
NM_003638.2 | NP_003629.2 | 17 - 18 | 2332 | 142 | |
XM_011519752.2 | XP_011518054.1 | 17 - 18 | 2326 | 142 | |
GenBank mRNA | AK297740.1 | - | 16 - 17 | 1799 | 142 |
L36531.1 | - | 17 - 18 | 1654 | 142 |