Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CAMK2D
Entrez Gene ID
817
Gene Name
calcium/calmodulin dependent protein kinase II delta
Gene Aliases
CAMKD
Chromosome Location
Chr.4: 113451032 - 113761927 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001221.3 NP_001212.2 5 - 6 1198 126
NM_001321566.1 NP_001308495.1 5 - 6 1198 126
NM_001321569.1 NP_001308498.1 5 - 6 1198 126
NM_001321570.1 NP_001308499.1 5 - 6 1198 126
NM_001321573.1 NP_001308502.1 5 - 6 1198 126
NM_001321574.1 NP_001308503.1 5 - 6 1198 126
NM_001321576.1 NP_001308505.1 5 - 6 1198 126
NM_001321579.1 NP_001308508.1 5 - 6 1198 126
NM_001321580.1 NP_001308509.1 5 - 6 1198 126
NM_001321581.1 NP_001308510.1 6 - 7 1299 126
NM_001321582.1 NP_001308511.1 5 - 6 1198 126
NM_001321584.1 NP_001308513.1 5 - 6 1198 126
NM_001321585.1 NP_001308514.1 6 - 7 1292 126
NM_001321586.1 NP_001308515.1 5 - 6 1198 126
NM_001321587.1 NP_001308516.1 5 - 6 1198 126
NM_001321588.1 NP_001308517.1 5 - 6 1198 126
NM_001321589.1 NP_001308518.1 5 - 6 1198 126
NM_001321590.1 NP_001308519.1 5 - 6 1198 126
NM_001321591.1 NP_001308520.1 5 - 6 1198 126
NM_172114.1 NP_742112.1 5 - 6 1198 126
NM_172115.2 NP_742113.1 5 - 6 1198 126
NM_172127.2 NP_742125.1 5 - 6 1198 126
NM_172128.2 NP_742126.1 5 - 6 1198 126
NM_172129.1 NP_742127.1 5 - 6 1198 126
XM_005263253.3 XP_005263310.1 5 - 6 1016 126
XM_011532289.1 XP_011530591.1 5 - 6 1016 126
XM_011532290.1 XP_011530592.1 5 - 6 1016 126
XM_011532291.1 XP_011530593.1 5 - 6 1016 126
XM_017008675.1 XP_016864164.1 6 - 7 1138 126
XM_017008676.1 XP_016864165.1 5 - 6 374 126
XM_017008678.1 XP_016864167.1 5 - 6 1016 126
GenBank mRNA AB209288.1 - 5 - 6 834 126
AF071569.1 - 5 - 6 339 126
AJ252239.1 - 5 - 6 772 126
AK055642.1 - 5 - 6 716 126
AY987011.1 - 5 - 6 339 126
BC032784.1 - 5 - 6 401 126

View Full Product Details