The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001011537.2 | NP_001011537.2 | 5 - 6 | 661 | 99 |
NM_032288.6 | NP_115664.2 | 4 - 5 | 720 | 99 | |
NR_027840.1 | 4 - 5 | 831 | 99 | ||
GenBank mRNA | AJ344094.1 | - | 4 - 5 | 566 | 99 |
AK027672.1 | - | 4 - 5 | 604 | 99 | |
AK302546.1 | - | 4 - 5 | 831 | 99 | |
AK303462.1 | - | 5 - 6 | 661 | 99 | |
AK315019.1 | - | 4 - 5 | 569 | 99 | |
AK316308.1 | - | 4 - 5 | 461 | 99 | |
AL136558.1 | - | 4 - 5 | 573 | 99 | |
BC035006.2 | - | 4 - 5 | 723 | 99 | |
BC039734.1 | - | 4 - 5 | 630 | 99 | |
CR533502.1 | - | 4 - 5 | 501 | 99 |