The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015015.2 | NP_055830.1 | 22 - 23 | 3344 | 167 |
XM_005259521.3 | XP_005259578.2 | 23 - 24 | 3447 | 167 | |
XM_011527814.2 | XP_011526116.1 | 22 - 23 | 3266 | 167 | |
XM_011527816.2 | XP_011526118.1 | 12 - 13 | 2796 | 167 | |
XM_011527817.2 | XP_011526119.1 | 15 - 16 | 3164 | 167 | |
XM_017026503.1 | XP_016881992.1 | 23 - 24 | 3447 | 167 | |
GenBank mRNA | AB020683.2 | - | 22 - 23 | 3336 | 167 |
AK126854.1 | - | 16 - 17 | 2884 | 167 | |
AL133622.1 | - | 11 - 12 | 1720 | 167 | |
BC136611.1 | - | 20 - 21 | 3158 | 167 | |
BC144292.1 | - | 21 - 22 | 3260 | 167 |