The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136000.2 | NP_001129472.1 | 2 - 3 | 217 | 108 |
NM_001168237.1 | NP_001161709.1 | 2 - 3 | 504 | 108 | |
NM_005158.4 | NP_005149.4 | 2 - 3 | 217 | 108 | |
NM_007314.3 | NP_009298.1 | 2 - 3 | 504 | 108 | |
GenBank mRNA | AK309549.1 | - | 2 - 3 | 204 | 108 |
BC065912.1 | - | 2 - 3 | 184 | 108 | |
BX538317.1 | - | 2 - 3 | 256 | 108 | |
DA596675.1 | - | 2 - 3 | 204 | 108 | |
FJ542284.1 | - | 2 - 3 | 217 | 108 | |
M35296.1 | - | 2 - 3 | 421 | 108 |