The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134492.1 | NP_001127964.1 | 3 - 4 | 836 | 152 |
NM_012262.3 | NP_036394.1 | 3 - 4 | 836 | 152 | |
GenBank mRNA | AB007917.1 | - | 3 - 4 | 733 | 152 |
AB024568.1 | - | 3 - 4 | 805 | 152 | |
BC025384.1 | - | 3 - 4 | 841 | 152 | |
BC025990.1 | - | 3 - 4 | 841 | 152 | |
BC108735.1 | - | 3 - 4 | 781 | 152 |