Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLCZ1
Entrez Gene ID
89869
Gene Name
phospholipase C zeta 1
Gene Aliases
NYD-SP27, PLC-zeta-1, PLCzeta
Chromosome Location
Chr.12: 18645529 - 18738059 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033123.3 NP_149114.2 11 - 12 1562 111
NR_073075.1 11 - 12 1565 111
XM_005253519.2 XP_005253576.1 10 - 11 1678 111
XM_011520883.1 XP_011519185.1 8 - 9 968 111
XM_017020176.1 XP_016875665.1 10 - 11 1678 111
XM_017020177.1 XP_016875666.1 11 - 12 1555 111
XM_017020178.1 XP_016875667.1 9 - 10 1528 111
XM_017020179.1 XP_016875668.1 10 - 11 1673 111
XM_017020180.1 XP_016875669.1 10 - 11 1567 111
XM_017020181.1 XP_016875670.1 10 - 11 1632 111
XM_017020182.1 XP_016875671.1 6 - 7 1331 111
XM_017020183.1 XP_016875672.1 7 - 8 1095 111
XM_017020184.1 XP_016875673.1 7 - 8 1023 111
XM_017020185.1 XP_016875674.1 6 - 7 908 111
XM_017020186.1 XP_016875675.1 8 - 9 930 111
GenBank mRNA AF532185.1 - 11 - 12 1487 111
AK097555.1 - 7 - 8 910 111
AK097725.1 - 7 - 8 1028 111
AK292279.1 - 11 - 12 1560 111
AK314516.1 - 11 - 12 1519 111
AY035866.1 - 11 - 12 1524 111
BC125067.1 - 7 - 8 815 111

View Full Product Details